Assay Details
| Gene Symbol: Hsp83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274433.1 | NP_001261362.1 | 2 | 2839 | 153 | |
| NM_079175.4 | NP_523899.1 | 2 | 2592 | 153 | ||
| GenBank mRNA | AY122080.1 | - | 2 | 2781 | 153 | |
More Information
Set Membership: |
|

Hsp83
Genomic Map