Assay Details
| Gene Symbol: mRpL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079560.5 | NP_524284.1 | 2 | 866 | 86 | |
| GenBank mRNA | AY061413.1 | - | 2 | 830 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mRpL19| Gene Symbol: mRpL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079560.5 | NP_524284.1 | 2 | 866 | 86 | |
| GenBank mRNA | AY061413.1 | - | 2 | 830 | 86 | |
Set Membership: |
|