Assay Details
| Gene Symbol: 5-HT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276202.1 | NP_001263131.1 | 3 | 3341 | 130 | |
| NM_079860.3 | NP_524599.1 | 2 | 3267 | 130 | ||
| GenBank mRNA | AY118491.1 | - | 2 | 3239 | 130 | |
More Information
Set Membership: |
|

5-HT7
Genomic Map