Assay Details
Gene Symbol: mEFTu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169673.2 | NP_001163144.1 | 1 | 1671 | 98 | |
NM_080013.4 | NP_524752.1 | 2 | 1615 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
