Assay Details
| Gene Symbol: CG12728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297989.1 | NP_001284918.1 | 5 | 1354 | 101 | |
| NM_132052.4 | NP_572280.2 | 5 | 1354 | 101 | ||
| GenBank mRNA | BT023834.1 | - | 5 | 1325 | 101 | |
More Information
Set Membership: |
|

CG12728
Genomic Map