Assay Details
Gene Symbol: Spt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133020.4 | NP_573248.2 | 3 | 1173 | 122 | |
GenBank mRNA | AY118561.1 | - | 3 | 1149 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
