Assay Details
| Gene Symbol: Cpr23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134874.4 | NP_608718.1 | 2 | 981 | 117 | |
| GenBank mRNA | AY071302.1 | - | 2 | 978 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpr23B| Gene Symbol: Cpr23B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134874.4 | NP_608718.1 | 2 | 981 | 117 | |
| GenBank mRNA | AY071302.1 | - | 2 | 978 | 117 | |
Set Membership: |
|