Assay Details
Gene Symbol: CG9107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135137.4 | NP_608981.1 | 2 | 858 | 56 | |
NM_164673.3 | NP_723144.1 | 2 | 858 | 56 | ||
GenBank mRNA | AY061376.1 | - | 2 | 846 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
