Assay Details
| Gene Symbol: Obp44a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299257.1 | NP_001286186.1 | 2 | 800 | 82 | |
| NM_136514.4 | NP_610358.1 | 2 | 572 | 82 | ||
| GenBank mRNA | AY070662.1 | - | 2 | 569 | 82 | |
More Information
Set Membership: |
|

Obp44a
Genomic Map