Assay Details
Gene Symbol: SkpB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136885.4 | NP_610729.1 | 2 | 712 | 78 | |
GenBank mRNA | BT023253.1 | - | 1 | 469 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
