Assay Details
| Gene Symbol: Mip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140714.4 | NP_648971.1 | 2 | 1160 | 136 | |
| GenBank mRNA | AY118306.1 | - | 2 | 1145 | 136 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mip| Gene Symbol: Mip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140714.4 | NP_648971.1 | 2 | 1160 | 136 | |
| GenBank mRNA | AY118306.1 | - | 2 | 1145 | 136 | |
Set Membership: |
|