Assay Details
Gene Symbol: RpL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275096.1 | NP_001262025.1 | 2 | 402 | 95 | |
NM_001316443.1 | NP_001303372.1 | 2 | 398 | 95 | ||
NM_140813.3 | NP_649070.1 | 2 | 398 | 95 | ||
GenBank mRNA | AY071133.1 | - | 2 | 426 | 95 | |
KX532097.1 | - | 1 | 348 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
