Assay Details
Gene Symbol: Rcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140987.3 | NP_649244.3 | 3 | 2109 | 145 | |
GenBank mRNA | AY075507.1 | - | 3 | 2106 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
