Assay Details
| Gene Symbol: Cpr78Cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275215.1 | NP_001262144.1 | 2 | 2062 | 101 | |
| NM_141042.2 | NP_649299.2 | 2 | 1158 | 101 | ||
| GenBank mRNA | AY075480.1 | - | 2 | 1153 | 101 | |
More Information
Set Membership: |
|

Cpr78Cb
Genomic Map