Assay Details
| Gene Symbol: SmD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141388.4 | NP_649645.1 | 2 | 648 | 121 | |
| GenBank mRNA | AY069315.1 | - | 2 | 606 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SmD2| Gene Symbol: SmD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141388.4 | NP_649645.1 | 2 | 648 | 121 | |
| GenBank mRNA | AY069315.1 | - | 2 | 606 | 121 | |
Set Membership: |
|