Assay Details
| Gene Symbol: SdhC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275543.1 | NP_001262472.1 | 2 | 581 | 69 | |
| NM_141790.3 | NP_650047.3 | 2 | 581 | 69 | ||
| GenBank mRNA | AY071728.1 | - | 2 | 582 | 69 | |
More Information
Set Membership: |
|

SdhC
Genomic Map