Assay Details
Gene Symbol: Obp99c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276148.1 | NP_001263077.1 | 2 | 361 | 80 | |
NM_143454.3 | NP_651711.1 | 2 | 375 | 80 | ||
GenBank mRNA | AY070983.1 | - | 2 | 370 | 80 | |
KX532148.1 | - | 2 | 314 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
