Assay Details
Gene Symbol: CG7950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143513.3 | NP_651770.1 | 2 | 1055 | 95 | |
NM_170464.3 | NP_733343.1 | 2 | 1079 | 95 | ||
GenBank mRNA | BT003292.1 | - | 2 | 1059 | 93 |
Gene Symbol: Rpp14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104507.3 | NP_001097977.1 | 2 | 1055 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
