Assay Details
| Gene Symbol: lwr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164385.2 | NP_722637.1 | 2 | 1171 | 142 | |
| GenBank mRNA | AY060944.1 | - | 2 | 1132 | 142 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lwr| Gene Symbol: lwr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164385.2 | NP_722637.1 | 2 | 1171 | 142 | |
| GenBank mRNA | AY060944.1 | - | 2 | 1132 | 142 | |
Set Membership: |
|