Assay Details
Gene Symbol: PIG-M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137756.4 | NP_611600.1 | 3 | 2010 | 56 | |
NM_166470.3 | NP_726100.1 | 2 | 2071 | 56 | ||
GenBank mRNA | AY047519.1 | - | 3 | 1990 | 56 |
Gene Symbol: CG42379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144262.2 | NP_001137734.1 | 3 | 2010 | 56 | |
NM_001144263.2 | NP_001137735.1 | 2 | 2071 | 56 |
Gene Symbol: CG42380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144264.2 | NP_001137736.1 | 3 | 2010 | 56 | |
NM_001144265.2 | NP_001137737.1 | 2 | 2071 | 56 |
Gene Symbol: CG42381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144261.2 | NP_001137733.1 | 3 | 2010 | 56 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
