Assay Details
| Gene Symbol: Act5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014725.2 | NP_001014725.1 | 2 | 1514 | 125 | |
| NM_001014726.2 | NP_001014726.1 | 2 | 1514 | 125 | ||
| NM_001297986.1 | NP_001284915.1 | 2 | 1514 | 125 | ||
| NM_078497.4 | NP_511052.1 | 2 | 1476 | 125 | ||
| NM_167053.2 | NP_727048.1 | 2 | 1514 | 125 | ||
| GenBank mRNA | AY089562.1 | - | 2 | 1508 | 125 | |
More Information
Set Membership: |
|

Act5C
Genomic Map