Assay Details
Gene Symbol: UbcE2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132210.3 | NP_572438.1 | 2 | 2113 | 163 | |
NM_167145.2 | NP_727233.1 | 2 | 2250 | 163 | ||
NM_167146.2 | NP_727234.1 | 2 | 2149 | 163 | ||
GenBank mRNA | AY061237.1 | - | 2 | 2092 | 163 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
