Assay Details
| Gene Symbol: RpS10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258829.2 | NP_001245758.1 | 2 | 715 | 155 | |
| NM_001298516.1 | NP_001285445.1 | 1 | 557 | 155 | ||
| NM_134480.4 | NP_608324.1 | 3 | 644 | 155 | ||
| NM_167668.2 | NP_728273.1 | 3 | 649 | 155 | ||
| GenBank mRNA | AY070896.1 | - | 903 | 155 | ||
More Information
Set Membership: |
|

RpS10b
Genomic Map