Assay Details
| Gene Symbol: caps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043139.4 | NP_001036604.2 | 5 | 1248 | 81 | |
| NM_001316462.1 | NP_001303391.1 | 5 | 1248 | 81 | ||
| NM_079332.4 | NP_524056.3 | 5 | 1248 | 81 | ||
| NM_168540.3 | NP_729880.2 | 4 | 1035 | 81 | ||
| GenBank mRNA | BT016006.1 | - | 4 | 1033 | 81 | |
More Information
Set Membership: |
|

caps
Genomic Map