Assay Details
Gene Symbol: Ndfip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168743.3 | NP_730291.1 | 2 | 1868 | 160 | |
GenBank mRNA | AY069849.1 | - | 2 | 1839 | 160 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
