Assay Details
| Gene Symbol: form3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014571.1 | NP_001014571.1 | 3 - 4 | 1163 | 62 | |
| NM_001104057.1 | NP_001097527.1 | 3 - 4 | 1163 | 62 | ||
| GenBank mRNA | AB061681.1 | - | 3 - 4 | 1162 | 62 | |
More Information
Set Membership: |
|

form3
Genomic Map