Assay Details
| Gene Symbol: form3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014571.1 | NP_001014571.1 | 4 - 5 | 1202 | 82 | |
| GenBank mRNA | AB061681.1 | - | 4 - 5 | 1201 | 82 | |
More Information
Set Membership: |
|

form3
Genomic Map