Assay Details
| Gene Symbol: Obp47b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136825.2 | NP_610669.1 | 1 - 2 | 112 | 174 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obp47b| Gene Symbol: Obp47b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136825.2 | NP_610669.1 | 1 - 2 | 112 | 174 | |
Set Membership: |
|