Assay Details
Gene Symbol: Rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_140901.3 | NP_649158.1 | 2 | 2941 | 157 | |
GenBank mRNA | AF132182.1 | - | 2 | 2886 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
