Assay Details
| Gene Symbol: Rpp14b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143512.2 | NP_651769.1 | 2 | 466 | 82 | |
| GenBank mRNA | BT120229.1 | - | 2 | 467 | 82 | |
| KX531530.1 | - | 2 | 287 | 82 | ||
More Information
Set Membership: |
|

Rpp14b
Genomic Map