Assay Details
Gene Symbol: RhoGAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103385.2 | NP_001096855.1 | 4 - 5 | 1841 | 77 | |
NM_001103386.2 | NP_001096856.1 | 4 - 5 | 1841 | 77 | ||
NM_001169155.1 | NP_001162626.1 | 3 - 4 | 2064 | 77 | ||
NM_001297797.1 | NP_001284726.1 | 3 - 4 | 1634 | 77 | ||
GenBank mRNA | BT024449.1 | - | 4 - 5 | 1803 | 77 | |
BT081989.1 | - | 3 - 4 | 2064 | 77 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
