Assay Details
Gene Symbol: RhoGAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103385.2 | NP_001096855.1 | 10 - 11 | 3769 | 73 | |
NM_001103386.2 | NP_001096856.1 | 10 - 11 | 3769 | 73 | ||
NM_001169155.1 | NP_001162626.1 | 8 - 9 | 3971 | 73 | ||
NM_001297795.1 | NP_001284724.1 | 10 - 11 | 3770 | 73 | ||
NM_001297796.1 | NP_001284725.1 | 10 - 11 | 3776 | 73 | ||
NM_001297797.1 | NP_001284726.1 | 8 - 9 | 3541 | 73 | ||
GenBank mRNA | BT024449.1 | - | 10 - 11 | 3731 | 73 | |
BT081989.1 | - | 8 - 9 | 3971 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
