Assay Details
| Gene Symbol: CG17528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042997.2 | NP_001036462.1 | 2 - 3 | 1069 | 75 | |
| NM_001042998.2 | NP_001036463.1 | 2 - 3 | 863 | 75 | ||
More Information
Set Membership: |
|

CG17528
Genomic Map