Assay Details
| Gene Symbol: Coa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042858.2 | NP_001036323.1 | 1 - 2 | 191 | 76 | |
| NM_001042859.2 | NP_001036324.1 | 1 - 2 | 191 | 76 | ||
| NM_001298612.1 | NP_001285541.1 | 1 - 2 | 191 | 76 | ||
| GenBank mRNA | AY122232.1 | - | 1 - 2 | 175 | 76 | |
More Information
Set Membership: |
|

Coa7
Genomic Map