Assay Details
Gene Symbol: RpL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057855.6 | NP_477203.1 | 3 | 121 | 140 | |
NM_166437.4 | NP_726053.1 | 2 | 181 | 140 | ||
NM_166438.4 | NP_726054.1 | 2 | 96 | 140 | ||
GenBank mRNA | KX531539.1 | - | 1 | 68 | 140 | |
U40226.1 | - | 2 | 97 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
