Assay Details
Gene Symbol: RpL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042975.2 | NP_001036440.1 | 2 | 242 | 61 | |
NM_001273771.1 | NP_001260700.1 | 2 | 242 | 61 | ||
GenBank mRNA | AY075491.1 | - | 2 | 241 | 61 | |
KX671630.1 | - | 1 | 161 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
