Assay Details
Gene Symbol: Tim23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015387.3 | NP_001015387.1 | 2 - 3 | 529 | 66 | |
GenBank mRNA | AY119236.1 | - | 2 - 3 | 556 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
