Assay Details
| Gene Symbol: Brca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138081.4 | NP_611925.2 | 1 - 2 | 877 | 104 | |
| GenBank mRNA | BT023811.1 | - | 1 - 2 | 784 | 104 | |
More Information
Set Membership: |
|

Brca2
Genomic Map