Assay Details
Gene Symbol: CG12567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015383.2 | NP_001015383.3 | 2 - 3 | 210 | 93 | |
NM_001015384.3 | NP_001015384.1 | 2 - 3 | 469 | 93 | ||
NM_001170362.1 | NP_001163833.1 | 2 - 3 | 200 | 93 | ||
NM_001300704.1 | NP_001287633.1 | 2 - 3 | 232 | 93 | ||
GenBank mRNA | AY071204.1 | - | 2 - 3 | 210 | 93 | |
BT083411.1 | - | 2 - 3 | 464 | 93 | ||
BT128756.1 | - | 2 - 3 | 200 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
