Assay Details
| Gene Symbol: CG12567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015383.2 | NP_001015383.3 | 3 - 4 | 863 | 69 | |
| NM_001015384.3 | NP_001015384.1 | 3 - 4 | 1122 | 69 | ||
| NM_001110861.3 | NP_001104331.1 | 3 - 4 | 1112 | 69 | ||
| NM_001170362.1 | NP_001163833.1 | 3 - 4 | 853 | 69 | ||
| NM_001170363.2 | NP_001163834.1 | 2 - 3 | 1174 | 69 | ||
| NM_001300704.1 | NP_001287633.1 | 3 - 4 | 885 | 69 | ||
| GenBank mRNA | AY071204.1 | - | 3 - 4 | 863 | 69 | |
| BT083411.1 | - | 3 - 4 | 1117 | 69 | ||
| BT128756.1 | - | 3 - 4 | 853 | 69 | ||
More Information
Set Membership: |
|

CG12567
Genomic Map