Assay Details
| Gene Symbol: Tim17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015401.3 | NP_001015401.1 | 1 - 2 | 313 | 100 | |
| NM_001300712.1 | NP_001287641.1 | 1 - 2 | 239 | 100 | ||
More Information
Set Membership: |
|

Tim17b
Genomic Map