Assay Details
Gene Symbol: Gprk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042973.3 | NP_001036438.1 | 11 - 12 | 1512 | 97 | |
NM_001169580.2 | NP_001163051.1 | 12 - 13 | 1427 | 97 | ||
NM_001299206.1 | NP_001286135.1 | 11 - 12 | 1512 | 97 | ||
GenBank mRNA | BT022770.1 | - | 11 - 12 | 1445 | 97 | |
BT022801.1 | - | 11 - 12 | 1487 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
