Assay Details
Gene Symbol: Gprk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042973.3 | NP_001036438.1 | 6 - 7 | 679 | 127 | |
NM_001169580.2 | NP_001163051.1 | 7 - 8 | 594 | 127 | ||
NM_001299206.1 | NP_001286135.1 | 6 - 7 | 679 | 127 | ||
GenBank mRNA | BT022770.1 | - | 6 - 7 | 612 | 127 | |
BT022801.1 | - | 6 - 7 | 654 | 127 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
