Assay Details
Gene Symbol: mRpS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043187.2 | NP_001036652.1 | 2 - 3 | 299 | 153 | |
NM_001043188.3 | NP_001036653.1 | 1 - 2 | 256 | 153 | ||
GenBank mRNA | AY071397.1 | - | 1 - 2 | 254 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
