Assay Details
| Gene Symbol: kdm5bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002166.2 | NP_001002166.2 | 8 - 9 | 1214 | 75 | |
| XM_005167024.4 | XP_005167081.1 | 9 - 10 | 1892 | 75 | ||
| XM_009303989.3 | XP_009302264.1 | 9 - 10 | 1892 | 75 | ||
| GenBank mRNA | BC071280.1 | - | 8 - 9 | 1147 | 75 | |
More Information
Set Membership: |
|

kdm5bb
Genomic Map