Assay Details
| Gene Symbol: dedd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002639.1 | NP_001002639.1 | 3 - 4 | 940 | 84 | |
| XM_005166826.4 | XP_005166883.1 | 3 - 4 | 993 | 84 | ||
| GenBank mRNA | BC075935.1 | - | 3 - 4 | 940 | 84 | |
More Information
Set Membership: |
|

dedd
Genomic Map