Assay Details
Gene Symbol: eci2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002645.3 | NP_001002645.3 | 5 - 6 | 677 | 82 | |
XM_021470136.1 | XP_021325811.1 | 5 - 6 | 588 | 82 | ||
XM_021470137.1 | XP_021325812.1 | 5 - 6 | 667 | 82 | ||
GenBank mRNA | BC076531.1 | - | 5 - 6 | 604 | 82 | |
BC155309.1 | - | 8 - 9 | 821 | 82 | ||
EH435933.1 | - | 5 - 6 | 629 | 82 | ||
EH589679.1 | - | 5 - 6 | 623 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
