Assay Details
| Gene Symbol: get1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003413.1 | NP_001003413.1 | 1 - 2 | 94 | 84 | |
| GenBank mRNA | AY648739.1 | - | 1 - 2 | 94 | 84 | |
| BC085456.1 | - | 1 - 2 | 92 | 84 | ||
| BC150353.1 | - | 1 - 2 | 94 | 84 | ||
More Information
Set Membership: |
|

get1
Genomic Map