Assay Details
| Gene Symbol: camk2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003602.2 | NP_001003602.1 | 12 - 13 | 1153 | 96 | |
| XM_005166561.4 | XP_005166618.1 | 12 - 13 | 1189 | 96 | ||
| XM_009303484.3 | XP_009301759.1 | 12 - 13 | 1191 | 96 | ||
| GenBank mRNA | BC077143.1 | - | 12 - 13 | 1121 | 96 | |
| Gene Symbol: camk2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CK687938.1 | - | 9 - 10 | 711 | 96 | |
More Information
Set Membership: |
|

camk2d1
Genomic Map