Assay Details
| Gene Symbol: camk2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003602.2 | NP_001003602.1 | 13 - 14 | 1186 | 89 | |
| XM_005166561.4 | XP_005166618.1 | 13 - 14 | 1222 | 89 | ||
| XM_009303484.3 | XP_009301759.1 | 13 - 14 | 1224 | 89 | ||
| GenBank mRNA | BC077143.1 | - | 13 - 14 | 1154 | 89 | |
| Gene Symbol: camk2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CK687938.1 | - | 10 - 11 | 744 | 89 | |
More Information
Set Membership: |
|

camk2d1
Genomic Map